Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-pentafluoro-L-phenylalanine 97.0+%, TCI America™
SDP

Catalog No. F09001G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F09001G 1 g
F0900200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F09001G Supplier TCI America Supplier No. F09001G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 205526-32-5
Molecular Formula C24H16F5NO4
Molecular Weight (g/mol) 477.39
MDL Number MFCD00270612
InChI Key DLOGILOIJKBYKA-KRWDZBQOSA-N
Synonym fmoc-pentafluoro-l-phenylalanine, fmoc-phe f5-oh, fmoc-l-pentafluorophenylalanine, fmoc-l-pentafluorophe, s-2-9h-fluoren-9-yl methoxy carbonyl amino-3-perfluorophenyl propanoic acid, fmoc-3-pentafluorophenyl-l-alanine, n-9h-fluoren-9-ylmethoxy carbonyl-pentafluoro-l-phenylalanine, n-alpha-9-fluorenylmethyloxycarbonyl-l-pentafluoro-phenylalanine, s-2-9h-fluoren-9-ylmethoxycarbonylamino-3-pentafluorophenyl-propionic acid, 2s-2-9h-fluoren-9-ylmethoxy carbonyl amino-3-pentafluorophenyl propanoic acid
PubChem CID 7020337
IUPAC Name (2S)-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)-3-(2,3,4,5,6-pentafluorophenyl)propanoic acid
SMILES OC(=O)[C@H](CC1=C(F)C(F)=C(F)C(F)=C1F)NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12

Specifications

Color White
Quantity 1 g
Formula Weight 477.39
Percent Purity ≥97.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-pentafluoro-L-phenylalanine
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.