Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Acetoacetylmorpholine 98.0+%, TCI America™
SDP

Catalog No. A0726500G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A0726500G 500 g
A072625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A0726500G Supplier TCI America Supplier No. A0726500G

Chemical Identifiers

CAS 16695-54-8
Molecular Formula C8H13NO3
Molecular Weight (g/mol) 171.20
MDL Number MFCD00059774
InChI Key RKPILPRSNWEZJV-UHFFFAOYSA-N
PubChem CID 85550
IUPAC Name 1-(morpholin-4-yl)butane-1,3-dione
SMILES CC(=O)CC(=O)N1CCOCC1

Specifications

Melting Point 68°C
Color White
Boiling Point 140°C
Quantity 500 g
Formula Weight 171.20
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material N-Acetoacetylmorpholine
EINECSNumber (9)-1635
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.