Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Acetyl-5-hydroxytryptamine, Thermo Scientific™

Catalog No. AC226930010
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC226930010 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AC226930010 Supplier Thermo Scientific Chemicals Supplier No. 226930010
Add to Cart
This item is not returnable. View return policy

Specifications

Chemical Name or Material N-Acetyl-5-hydroxytryptamine
CAS 1210-83-9
Synonym N-Acetylserotonin
Quantity 1 g
Melting Point 119°C to 123°C
Shipping Condition No special requirements
Product Line Life science
Molecular Formula C12 H14 N2 O2
Physical Form Powder
Color White to Light-yellow
Purity ≥97.5%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.