Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals N-Acetyl-Arg-Glu-Lys-Arg-7-amino-4-(trifluoromethyl)coumarin

Catalog No. AAJ64106MA
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ64106MA 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAJ64106MA Supplier Thermo Scientific Chemicals Supplier No. J64106MA
Get 15% Off Thermo Scientific Reagents
Use promo code REAGENTS15
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

C35H51F3N12O9

N-Acetyl-Arg-Glu-Lys-Arg-7-amino-4-(trifluoromethyl)coumarin is employed as a fluorogenic cathepsin D substrate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Quantity 10 mg
Description White
Content And Storage 2°C to 8°C
For Use With (Application) A fluorogenic substrate suitable for the assay of furin
Molecular Formula C35H51F3N12O9
Formula Weight 840.80g/mol
Synonym Ac-REKR-AFC; Ac-Arg-Glu-Lys-Arg-AFC
Sensitivity Light Sensitive
Solubility Soluble in DMSO or DMF
Product Type Signal Transduction Reagent
Form Solid
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.