Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals N-Acetyl-D-galactosamine

Catalog No. AAJ6609503
Change view
Click to view available options
Quantity:
500 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6609503 1 g
AAJ66095ME 500 mg
AAJ6609506 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAJ6609503 Supplier Thermo Scientific Chemicals Supplier No. J6609503
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 1811-31-0 | C8H15NO6 | 221.21 g/mol

N-Acetyl-D-galactosamine is the initial O-linked sugar to many serine and threonine residues in protein glycosylation.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-Acetyl-D-galactosamine is the initial O-linked sugar to many serine and threonine residues in protein glycosylation.

Solubility
Soluble in water at 50mg/ml

Notes
Light Sensitive & Hygroscopic. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1811-31-0
Molecular Formula C8H15NO6
Molecular Weight (g/mol) 221.21
MDL Number MFCD00136044
InChI Key OVRNDRQMDRJTHS-KEWYIRBNSA-N
Synonym dsstox_cid_28637, dsstox_rid_82907, dsstox_gsid_48711, n-acetyl-, a-d-galactosamine
PubChem CID 60196345
SMILES CC(=O)N[C@H]1C(O)O[C@H](CO)[C@H](O)[C@@H]1O

Specifications

Melting Point 160°C
Color White
Quantity 1 g
Beilstein 2331340
Solubility Information Soluble in water at 50mg/ml
IUPAC Name N-[(2R,3S,4S,5S)-3,4,5,6-tetrahydroxy-1-oxohexan-2-yl]acetamide
Formula Weight 221.21
Physical Form Solid
Chemical Name or Material N-Acetyl-D-galactosamine
EINECSNumber 217-321-9
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.