Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abcam N-Acetyl-D-glucosamine, 221.21 Da, >98% Purity, 25 g

Catalog No. 502704821
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-270-4821 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-270-4821 Supplier Abcam Supplier No. AB14622725G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Monosaccharide derivative used to characterize N-acetyl-β-D-hexoaminidase(s), with >98% purity.

  • Molecular Weight: 221.21 Da
  • Purity: >98%
  • Applications: Used to identify, differentiate and characterize N-acetyl-β-D-hexoaminidase(s)
  • Composition: 100% N-Acetyl-D-glucosamine
  • Storage: Store at +4°C
  • Shipping: Ambient - Can Ship with Ice

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Target Molecule N-Acetyl-β-D-hexoaminidase
Substrate Properties Chemical Substrate
Substrate N-Acetyl-β-D-hexoaminidase
Shipping Condition Ambient
Buffer 100% N-Acetyl-D-glucosamine
Content And Storage Store at 4°C.
Formulation 100% N-Acetyl-D-glucosamine
Quantity 25 g
For Use With (Application) Identify, differentiate and characterize N-acetyl-β-D-hexoaminidase(s)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.