Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Acetyl-D-phenylalanine, 95%, Thermo Scientific™

Catalog No. AAH6338206
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6338206 5 g
AAH6338214 25 g
AAH6338222 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH6338206 Supplier Thermo Scientific Chemicals Supplier No. H6338206

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10172-89-1
Molecular Formula C11H13NO3
Molecular Weight (g/mol) 207.23
MDL Number MFCD00002664
InChI Key CBQJSKKFNMDLON-SNVBAGLBSA-N
Synonym n-acetyl-d-phenylalanine, acetyl-d-phenylalanine, ac-d-phe-oh, n-acetyl-3-phenyl-d-alanine, n-acetylphenylalanine, d, d-phenylalanine, n-acetyl, r-2-acetamido-3-phenylpropanoic acid, 2r-2-acetamido-3-phenylpropanoic acid, n-acetylphenylalanine #, n-ac-d-phe-oh
PubChem CID 101184
ChEBI CHEBI:28203
IUPAC Name (2R)-2-acetamido-3-phenylpropanoic acid
SMILES CC(=O)N[C@H](CC1=CC=CC=C1)C(O)=O

Specifications

Melting Point 165°C to 169°C
Color White
Beilstein 2213852
Quantity 5 g
Formula Weight 207.23
Percent Purity 95%
Physical Form Solid
Chemical Name or Material N-Acetyl-D-phenylalanine
EINECSNumber 233-447-7
TSCA No
storageNote1 -30° to -10°C
Recommended Storage Store at -20°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.