Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Acetyl-D-tryptophan 98.0+%, TCI America™
SDP

Catalog No. A01191G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A01191G 1 g
A01195G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A01191G Supplier TCI America Supplier No. A01191G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 2280-01-5
Molecular Formula C13H14N2O3
Molecular Weight (g/mol) 246.27
MDL Number MFCD00065021
InChI Key DZTHIGRZJZPRDV-GFCCVEGCSA-N
Synonym Ac-D-Trp-OH
PubChem CID 439917
ChEBI CHEBI:16734
IUPAC Name (2R)-2-acetamido-3-(1H-indol-3-yl)propanoic acid
SMILES CC(=O)NC(CC1=CNC2=CC=CC=C21)C(=O)O

Specifications

Melting Point 186°C
Color White
Quantity 1 g
Formula Weight 246.27
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material N-Acetyl-D-tryptophan
EINECSNumber (9)-1601
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.