Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Acetyl-DL-2-aminobutyric Acid 97.0+%, TCI America™
SDP

Catalog No. A085325G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A085325G 25 g
A08535G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A085325G Supplier TCI America Supplier No. A085325G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 7211-57-6
Molecular Formula C6H11NO3
Molecular Weight (g/mol) 145.158
MDL Number MFCD00020430
InChI Key WZVZUKROCHDMDT-UHFFFAOYSA-N
Synonym 2-Acetamidobutyric Acid
PubChem CID 306107
IUPAC Name 2-acetamidobutanoic acid
SMILES CCC(C(=O)O)NC(=O)C

Specifications

Melting Point 132°C
Color White
Quantity 25 g
Formula Weight 145.16
Percent Purity ≥97.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material N-Acetyl-DL-2-aminobutyric Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.