Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Acetyl-DL-leucine 99.0+%, TCI America™
SDP

Catalog No. A009725G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A009725G 25 g
A00975G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A009725G Supplier TCI America Supplier No. A009725G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 99-15-0
Molecular Formula C8H15NO3
Molecular Weight (g/mol) 173.21
MDL Number MFCD00026498
InChI Key WXNXCEHXYPACJF-UHFFFAOYNA-N
Synonym n-acetyl-dl-leucine, acetylleucine, ac-dl-leu-oh, tanganil, l-leucine, n-acetyl, acetyl-dl-leucine, acetylleucine inn, dl-leucine, n-acetyl, 2-acetylamino-4-methylpentanoic acid
PubChem CID 1995
IUPAC Name 2-acetamido-4-methylpentanoic acid
SMILES CC(C)CC(NC(C)=O)C(O)=O

Specifications

Melting Point 160°C
Color White
Quantity 25 g
Formula Weight 173.21
Percent Purity ≥99.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material N-Acetyl-DL-leucine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.