Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Acetyl-L-aspartic acid 4-tert-butyl ester, 95%, Thermo Scientific™

Catalog No. AAH63933MD
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH63933MD 250 mg
AAH6393303 1 g
AAH6393306 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH63933MD Supplier Thermo Scientific Chemicals Supplier No. H63933MD

Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please to view product availability in your area.

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 117833-18-8
Molecular Formula C10H17NO5
Molecular Weight (g/mol) 231.248
MDL Number MFCD00236746
InChI Key NKNPTCBHHPHSEA-ZETCQYMHSA-N
Synonym ac-asp otbu-oh, s-2-acetamido-4-tert-butoxy-4-oxobutanoic acid, n-acetyl-l-aspartic acid 4-tert-butyl ester, acetyl-l-aspartic acid beta-tert-butyl ester, 2s-4-tert-butoxy-2-acetamido-4-oxobutanoic acid, ac-asp-otbu, s-3-acetamido-4-tert-butoxy-4-oxobutanoic acid, l-aspartic acid,n-acetyl-,4-1,1-dimethylethyl ester, l-aspartic acid, n-acetyl-, 4-1,1-dimethylethyl ester
PubChem CID 7019596
IUPAC Name (2S)-2-acetamido-4-[(2-methylpropan-2-yl)oxy]-4-oxobutanoic acid
SMILES CC(=O)NC(CC(=O)OC(C)(C)C)C(=O)O

Specifications

Quantity 250 mg
Formula Weight 231.25
Percent Purity 95%
Physical Form Solid
Chemical Name or Material N-Acetyl-L-aspartic acid 4-tert-butyl ester
TSCA No
storageNote1 2° to 8°C
Recommended Storage Keep cold
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.