Learn More
Enzo Life Sciences N-Acetyl-L-cysteine (5g). CAS: 616-91-1
Supplier: Enzo Life Sciences ALX105005G005
Derivative of cysteine wherein an acetyl group is attached to the amino group. Is used as a specific antidote in cases of acetaminophen overdose. Alternative name: LNAL, N-Acetyl-3-mercaptoalanine, L-alpha-Acetamido-beta-mercaptopropionic acid, NAC. Formula: C5H9NO3S. MW: 163.2. MI: 14: 88. Purity: ≥98% (TLC). Appearance: White powder. Solubility: Soluble in 5M hydrochloric acid (hot) (100mg/ml), methanol (50mg/ml), water or 100% ethanol; insoluble in chloroform or ether. Long Term Storage: +4°C.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.