Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Acetyl-L-glutamic acid 5-tert-butyl ester, 95%

Catalog No. AAH63871MD
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH63871MD 250 mg
AAH6387103 1 g
AAH6387106 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH63871MD Supplier Thermo Scientific Chemicals Supplier No. H63871MD

CAS: 84192-88-1 | C11H19NO5 | 245.28 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 84192-88-1
Molecular Formula C11H19NO5
Molecular Weight (g/mol) 245.28
MDL Number MFCD00236754
InChI Key FALCCLKESWGSNI-UHFFFAOYNA-N
Synonym ac-glu otbu-oh, s-2-acetamido-5-tert-butoxy-5-oxopentanoic acid, 2s-5-tert-butoxy-2-acetamido-5-oxopentanoic acid
PubChem CID 7019606
SMILES CC(=O)NC(CCC(=O)OC(C)(C)C)C(O)=O

Specifications

Color White
IUPAC Name (2S)-2-acetamido-5-[(2-methylpropan-2-yl)oxy]-5-oxopentanoic acid
Quantity 250 mg
Formula Weight 245.27
Percent Purity 95%
Physical Form Powder
Chemical Name or Material N-Acetyl-L-glutamic acid 5-tert-butyl ester
TSCA No
storageNote1 2° to 8°C
Recommended Storage Keep cold

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.