Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Acetyl-L-glutamic Acid 98.0+%, TCI America™
SDP

Catalog No. A0693250G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A0693250G 250 g
A069325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A0693250G Supplier TCI America Supplier No. A0693250G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 1188-37-0
Molecular Formula C7H11NO5
Molecular Weight (g/mol) 189.167
MDL Number MFCD00002802
InChI Key RFMMMVDNIPUKGG-YFKPBYRVSA-N
Synonym n-acetyl-l-glutamic acid, acetylglutamic acid, n-acetyl-l-glutamate, n-acetylglutamic acid, n-acetylglutamate, acetyl glutamic acid, acetyl-l-glutamic acid, l-glutamic acid, n-acetyl, l-n-acetylglutamic acid, ac-glu-oh
PubChem CID 70914
ChEBI CHEBI:17533
IUPAC Name (2S)-2-acetamidopentanedioic acid
SMILES CC(=O)NC(CCC(=O)O)C(=O)O

Specifications

Melting Point 198°C
Color White
Quantity 250 g
Formula Weight 189.17
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material N-Acetyl-L-glutamic Acid
EINECSNumber (9)-2029
RTECSNumber LZ9725000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.