Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals N-Acetyl-L-phenylalanine, 99%

Catalog No. AAB2381206
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2381206 5 g
AAB2381203 1 g
AAB2381214 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB2381206 Supplier Thermo Scientific Chemicals Supplier No. B2381206
Add to Cart

CAS: 2018-61-3 | C11H13NO3 | 207.23 g/mol

It is an essential amino acid produced for medical, feed, and nutritional applications such as in the preparation of aspartame. It is applied as an antidepressant.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an essential amino acid produced for medical, feed, and nutritional applications such as in the preparation of aspartame. It is applied as an antidepressant.

Solubility
Solubility in methanol (almost transparency). Soluble in acetone and ethanol (20 mg/ ml).

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2018-61-3
Molecular Formula C11H13NO3
Molecular Weight (g/mol) 207.23
MDL Number MFCD00063158
InChI Key CBQJSKKFNMDLON-JTQLQIEISA-N
Synonym n-acetyl-l-phenylalanine, ac-phe-oh, acetyl-l-phenylalanine, acetylphenylalanine, l-n-acetylphenylalanine, s-2-acetamido-3-phenylpropanoic acid, phenylalanine, n-acetyl, n-acetyl-3-phenyl-l-alanine, unii-np5bt39467, 2s-2-acetamido-3-phenylpropanoic acid
PubChem CID 74839
ChEBI CHEBI:16259
IUPAC Name (2S)-2-acetamido-3-phenylpropanoic acid
SMILES CC(=O)N[C@@H](CC1=CC=CC=C1)C(O)=O

Specifications

Melting Point 166°C to 170°C
Quantity 5 g
Formula Weight 207.23
Optical Rotation +40° (c=1 in Methanol)
Percent Purity 99%
Chemical Name or Material N-Acetyl-L-phenylalanine
EINECSNumber 217-959-8
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.