Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Acetyl-L-valine 98.0+%, TCI America™
SDP

Catalog No. A236625G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A236625G 25 g
A23665G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A236625G Supplier TCI America Supplier No. A236625G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 96-81-1
Molecular Formula C7H13NO3
Molecular Weight (g/mol) 159.19
MDL Number MFCD00066066
InChI Key IHYJTAOFMMMOPX-LURJTMIESA-N
Synonym n-acetyl-l-valine, ac-val-oh, l-valine, n-acetyl, s-2-acetamido-3-methylbutanoic acid, acetyl-l-valine, n-acetylvaline, 2s-2-acetamido-3-methylbutanoic acid, unii-u83p7h9hv3, acetylvaline, valine, n-acetyl-, l
PubChem CID 66789
IUPAC Name (2S)-2-acetamido-3-methylbutanoic acid
SMILES CC(C)[C@H](NC(C)=O)C(O)=O

Specifications

Melting Point 165°C
Color White
Quantity 25 g
Formula Weight 159.19
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material N-Acetyl-L-valine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.