Learn More
Enzo Life Sciences N-Acetyl-leukotriene E4 (50µg). CAS: 80115-95-3
SDP
Supplier:  Enzo Life Sciences BMLLEA0040050
Encompass_Preferred
Major bilary metabolite of cysteinyl leukotrienes in the rat. Approximately 100-fold less active than LTC4. Equiactive with LTE4 in guinea pig lung parenchyma assay. Alternative name: N-Acetyl-LTE4. Formulation: Solution in methanol/ammonium acetate buffer, 70:30. Purity: ≥98% (HPLC). Solubility: Soluble in ethanol. Long Term Storage: -80°C.
Catalog No. 50-201-1149
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.