Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals N-Acetyl-Tyr-Glu-Val-Asp-7-amino-4-methylcoumarin

Catalog No. AAJ65646LB0
Change view
Click to view available options
Quantity:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ65646LB0 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAJ65646LB0 Supplier Thermo Scientific Chemicals Supplier No. J65646LB0
Add to Cart

missing translation for 'validMainframeMsg'

 

A fluorogenic substrate for caspase 1

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Quantity 5 mg
Description White to off-white
Content And Storage -30°C to -10°C
For Use With (Application) A fluorogenic substrate for caspase 1
Molecular Formula C35H41N5O12
Formula Weight 723.72
Synonym Ac-YEVD-AMC; Caspase 1 (ICE) Substrate 1M fluorogenic
Sensitivity Light Sensitive
Product Type N-Acetyl-Tyr-Glu-Val-Asp-7-amino-4-methylcoumarin
Form Powder

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.