Learn More
Selleck Chemical LLC N-Acetylcysteine amide 5mg 38520-57-9
SDP
Supplier:  Selleck Chemical LLC S58045MG
Encompass_Preferred
N-acetylcysteine amide is a membrane penetrating antioxidant with anti-inflamatory activity through regulation of activation of NF-?B and HIF-1? as well as modulation of ROS. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-194-8175
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.