Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Acetylethylenediamine 95.0+%, TCI America™
SDP

Catalog No. A088725G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A088725G 25 g
A08875G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A088725G Supplier TCI America Supplier No. A088725G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 1001-53-2
Molecular Formula C4H10N2O
Molecular Weight (g/mol) 102.137
MDL Number MFCD00008163
InChI Key DAKZISABEDGGSV-UHFFFAOYSA-N
Synonym n-2-aminoethyl acetamide, n-acetylethylenediamine, acetamide, n-2-aminoethyl, n-2-amino-ethyl-acetamide, unii-fql33v3kzd, fql33v3kzd, acetylethylenediamine, 2-acetamidoethylamine, acetylethylene diamine, n-acetylethylendiamine
PubChem CID 66082
IUPAC Name N-(2-aminoethyl)acetamide
SMILES CC(=O)NCCN

Specifications

Color White-Yellow
Boiling Point 143°C
Quantity 25 g
UN Number 3259
Formula Weight 102.14
Percent Purity ≥95.0% (GC)
Physical Form Crystals
Chemical Name or Material N-Acetylethylenediamine
EINECSNumber (2)-0722
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.