Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abcam N-Acetylglucosamine BSA Glycoconjugate, Bovine, Native, Unconjugated, 50 μg

Catalog No. 502672441
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-267-2441 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-267-2441 Supplier Abcam Supplier No. AB24593250UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

High purity cow full length protein suitable for ELISA, WB and SDS-PAGE applications.

  • Species: Cow
  • Expression System: Native
  • Purity: >98%
  • Formulation: Liquid, PBS with 0.09% sodium azide, pH 7.4
  • Tested Applications: ELISA, SDS-PAGE, WB
  • Storage Conditions: Store at -20°C

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Target Molecule N-Acetylglucosamine, Bovine Serum Albumin
Detection Location In-Blot Detection
Label or Dye Unconjugated
Shipping Condition Blue Ice
Substrate Properties Protein-based Substrate
Buffer PBS, 0.09% sodium azide, pH7.4
Content And Storage Store at -20°C.
Form Liquid
Formulation PBS, 0.09% sodium azide, pH 7.4
Quantity 50 μg
For Use With (Application) SDS-PAGE, Western Blot, ELISA
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.