Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Acetylneuraminic acid, MP Biomedicals™

Catalog No. MP215385191 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP215385191 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MP215385191 Supplier MP Biomedicals, Inc Supplier No. 0215385191
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

N-Acetylneuraminic acid ≥98%, Off-White Powder

  • N-Acetylneuraminic acid (NANA, Neu5Ac) is a major component of glycoconjugates such as glycolipids, glycoproteins and proteoglycans (sialogylcoproteins) where it confers selective binding characteristics to the glycosylated component.
  • Neu5Ac is used to study its biochemistry, metabolism and uptake in vivo and in vitro. Neu5Ac is used in the development of nanocarriers.
  • Both sialic acid and neuraminic acid are loosely used to refer to conjugates of neuraminic acid. N-Acetylneuraminic acid is often found as the terminal sugar of cell surface glycoproteins.
  • Cell surface glycoproteins have important roles in cell recognition and interaction as well as in cell adhesion.
  • Cell surface glycoproteins have important roles in cell recognition and interaction as well as in cell adhesion.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 131-48-6
Molecular Formula C11H19NO9
Molecular Weight (g/mol) 309.27
MDL Number MFCD00006620
InChI Key SQVRNKJHWKZAKO-UHFFFAOYNA-N
Synonym Sialic Acid, NANA, Type IV, 5-Acetamido-3,5-dideoxy-D-glycero-D-galactononulosonic acid

Specifications

Melting Point 180°C to 190°C
Quantity 1 g
Beilstein 1716283
Solubility Information Soluble in water
Physical Form Powder
Grade >98%
storageNote1 +4°C
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.