Learn More
Abcam N-Acetylphytosphingosine, Sphingolipid Ceramide Derivative, >98% Purity, 5 mg
Description
- Molecular Weight: 359.5 Da
- Purity: >98%
- Chemical Class: Sphingolipid ceramide derivative
- Biological Activity: Induces caspase-dependent apoptosis
- Mechanism of Action: Potential NF-κB and JAK/STAT signaling inhibitor; acts as an anticancer agent
- Usage: Active in vitro and potentially in vivo
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Specifications
| Target Molecule | NF-κB, JAK/STAT |
| Shipping Condition | Ambient |
| Content And Storage | Store at -20°C. |
| Formulation | 100% N-[(2S,3S,4R)-1,3,4-trihydroxyoctadecan-2-yl]acetamide |
| Quantity | 5 mg |
| For Use With (Application) | Cancer research, Apoptosis, Signal transduction inhibition |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.