Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Acetylsulfanilyl chloride, 99%, ACROS Organics™

Click to view available options
Quantity:
100g
500g
2.5kg
Packaging:
Glass bottle
Plastic Bottle
121-60-8
C8H8ClNO3S
233.67
MFCD00007442
GRDXCFKBQWDAJH-UHFFFAOYSA-N
n-acetylsulfanilyl chloride,4-acetamidobenzene-1-sulfonyl chloride,dagenan chloride,4-acetylamino benzenesulfonyl chloride,4-acetamidophenylsulfonyl chloride,benzenesulfonyl chloride, 4-acetylamino,acetylsulfanilyl chloride,p-acetamidobenzenesulfonyl chloride,n-acetylsulphanilyl chloride,p-acetaminobenzenesulfonyl chloride
8481
4-acetamidobenzenesulfonyl chloride
CC(=O)NC1=CC=C(C=C1)S(=O)(=O)Cl
121-60-8
C8H8ClNO3S
233.67
MFCD00007442
GRDXCFKBQWDAJH-UHFFFAOYSA-N
n-acetylsulfanilyl chloride,4-acetamidobenzene-1-sulfonyl chloride,dagenan chloride,4-acetylamino benzenesulfonyl chloride,4-acetamidophenylsulfonyl chloride,benzenesulfonyl chloride, 4-acetylamino,acetylsulfanilyl chloride,p-acetamidobenzenesulfonyl chloride,n-acetylsulphanilyl chloride,p-acetaminobenzenesulfonyl chloride
8481
4-acetamidobenzenesulfonyl chloride
CC(=O)NC1=CC=C(C=C1)S(=O)(=O)Cl
Packaging Glass bottle
Flash Point >112°C
Infrared Spectrum Authentic
Quantity 100g
Melting Point 145.0°C to 148.0°C
Color Beige-Cream to White
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 98.5% min. (Argentometry)
Linear Formula 4-(CH3CONH)C6H4SO2Cl
Beilstein 14, 702
Fieser 01,3
Merck Index 15, 95
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in chloroform,ethylene dichloride,,alcohol,ether
Formula Weight 233.67
Physical Form Granular Crystalline Powder or Crystals
Percent Purity 99%
Chemical Name or Material N-Acetylsulfanilyl chloride, 99%
Show More Show Less

GHS H Statement
Causes severe skin burns and eye damage.

GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre

GHS Signal Word: Danger

EINECSNumber : 204-485-1

RTECSNumber : DB8837500

TSCA : TSCA