Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Acetyltryptamine 98.0+%, TCI America™
SDP

Catalog No. A2685100MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A2685100MG 100 mg
A268525MG 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A2685100MG Supplier TCI America Supplier No. A2685100MG

Chemical Identifiers

CAS 1016-47-3
Molecular Formula C12H14N2O
Molecular Weight (g/mol) 202.257
MDL Number MFCD00209910
InChI Key NVUGEQAEQJTCIX-UHFFFAOYSA-N
Synonym n-acetyltryptamine, n-2-1h-indol-3-yl ethyl acetamide, 3-2-n-acetylaminoethyl indole, acetamide, n-2-1h-indol-3-yl ethyl, chembl33171, n-2-1h-indol-3-yl-ethyl-acetamide, acetotryptamide, acetamide, n-2-indol-3-ylethyl
PubChem CID 70547
ChEBI CHEBI:55515
IUPAC Name N-[2-(1H-indol-3-yl)ethyl]acetamide
SMILES CC(=O)NCCC1=CNC2=CC=CC=C21

Specifications

Melting Point 76°C
Boiling Point 200°C
Quantity 100 mg
Formula Weight 202.26
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material N-Acetyltryptamine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.