Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-alpha-Benzoyl-L-arginine ethyl ester hydrochloride, 98+%

Catalog No. p-7024028
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1818103 1 g
AAA1818106 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1818103 Supplier Thermo Scientific Chemicals Supplier No. A1818103
Add to Cart

CAS: 2645-08-1 | C16H17ClN4O2S | 364.85 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2645-08-1
MDL Number MFCD00012579
ChEBI CHEBI:87675

Specifications

Melting Point 127°C to 131°C
Molecular Formula C16H17ClN4O2S
Beilstein 3781694
Synonym methylene green, basic green 5, c.i. basic green 5, c.i. basic green 5 van, methylene green van, unii-f1io2zp7bu, f1io2zp7bu, phenothiazin-5-ium, 3,7-bis dimethylamino-4-nitro-, chloride, 3,7-bis dimethylamino-4-nitrophenothiazin-5-ium chloride, 7-dimethylamino-n,n-dimethyl-4-nitrophenothiazin-3-iminium chloride
InChI Key YYGBVRCTHASBKD-UHFFFAOYSA-M
SMILES CN(C)C1=CC2=C(C=C1)N=C3C=CC(=[N+](C)C)C(=C3S2)[N+](=O)[O-].[Cl-]
IUPAC Name [7-(dimethylamino)-4-nitrophenothiazin-3-ylidene]-dimethylazanium;chloride
Quantity 1 g
Molecular Weight (g/mol) 364.85
PubChem CID 75889
Formula Weight 342.84
Optical Rotation −17° (c=2, Water)
Sensitivity Hygroscopic
Percent Purity ≥98%
Chemical Name or Material N-alpha-Benzoyl-L-arginine ethyl ester hydrochloride
Show More Show Less
EINECSNumber 220-157-0
TSCA No
Recommended Storage Store at -20°C; Store under Nitrogen

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.