Learn More
Thermo Scientific Chemicals N-alpha-Benzoyl-L-arginine p-nitroanilide hydrochloride
Description
N-alpha-Benzoyl-L-arginine p-nitroanilide hydrochloride is used as a chromogenic substrate for trypsin and other proteolytic enzymes.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
N-alpha-Benzoyl-L-arginine p-nitroanilide hydrochloride is used as a chromogenic substrate for trypsin and other proteolytic enzymes.
Solubility
Soluble in DMSO at 50mg/ml
Notes
Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Specifications
Specifications
| Melting Point | 223°C |
| Color | White to Yellow |
| Beilstein | 4081878 |
| Solubility Information | Soluble in DMSO at 50mg/ml |
| Quantity | 250 mg |
| Formula Weight | 434.9 |
| Physical Form | Powder |
| Chemical Name or Material | N-alpha-Benzoyl-L-arginine p-nitroanilide hydrochloride |
Safety and Handling
| EINECSNumber | 244-505-6 |
| TSCA | No |
| Recommended Storage | Store at -20°C |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.