Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Aminophthalimide 98.0+%, TCI America™
SDP

Catalog No. A084325G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A084325G 25 g
A08435G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A084325G Supplier TCI America Supplier No. A084325G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 1875-48-5
Molecular Formula C8H6N2O2
Molecular Weight (g/mol) 162.148
MDL Number MFCD00005895
InChI Key KSILMCDYDAKOJD-UHFFFAOYSA-N
Synonym n-aminophthalimide, 2-aminoisoindoline-1,3-dione, 2-amino-1h-isoindole-1,3 2h-dione, phthalimide, n-amino, hydrazine, 1,1-phthaloyl, phthalhydrazide van, 1h-isoindole-1,3 2h-dione, 2-amino, n-aminophthalamide, phthalocyclohydrazide van, ccris 5349
PubChem CID 74645
IUPAC Name 2-aminoisoindole-1,3-dione
SMILES C1=CC=C2C(=C1)C(=O)N(C2=O)N

Specifications

Color White-Yellow
Quantity 25 g
Formula Weight 162.15
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material N-Aminophthalimide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.