Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Benzyl-2-naphthylamine 98.0+%, TCI America™
SDP

Catalog No. B308825G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B308825G 25 g
B30885G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B308825G Supplier TCI America Supplier No. B308825G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 13672-18-9
Molecular Formula C17H15N
Molecular Weight (g/mol) 233.314
MDL Number MFCD00144848
InChI Key KBOITYDYBXNTMF-UHFFFAOYSA-N
PubChem CID 12487301
IUPAC Name N-benzylnaphthalen-2-amine
SMILES C1=CC=C(C=C1)CNC2=CC3=CC=CC=C3C=C2

Specifications

Melting Point 68°C
Boiling Point 190°C
Quantity 25 g
Formula Weight 233.31
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material N-Benzyl-2-naphthylamine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.