Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Benzyl-2-pyridineethylamine, Thermo Scientific™

Catalog No. AAH50257MD
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH50257MD 250 mg
AAH5025703 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH50257MD Supplier Thermo Scientific Chemicals Supplier No. H50257MD

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6312-25-0
Molecular Formula C14H16N2
Molecular Weight (g/mol) 212.296
MDL Number MFCD00089263
InChI Key WUDUNBALAGWKQB-UHFFFAOYSA-N
Synonym n-benzyl-2-pyridin-2-yl ethanamine, n-benzyl-2-pyridin-2-yl-ethanamine, benzyl 2-pyridin-2-yl ethyl amine, n-benzyl-2-pyridin-2-ylethylamine, benzyl 2-2-pyridyl ethyl amine, acmc-20ak6u, n-benzyl-2-pyridineethylamine, 2-2-benzylaminoethyl pyridine, benzyl-2-pyridin-2-yl-ethyl amine, n-phenylmethyl-2-pyridin-2-ylethanamine
PubChem CID 238354
IUPAC Name N-benzyl-2-pyridin-2-ylethanamine
SMILES C1=CC=C(C=C1)CNCCC2=CC=CC=N2

Specifications

Quantity 250 mg
UN Number UN2735
Formula Weight 212.3
Chemical Name or Material N-Benzyl-2-pyridineethylamine
Hazard Category Danger
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, N.O.S.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.