Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Benzyl-4-fluorobenzamide, 97%, Thermo Scientific™

Catalog No. AAH5762903
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5762903 1 g
AAH57629MD 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5762903 Supplier Thermo Scientific Chemicals Supplier No. H5762903

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
It is used in the microwave-assisted method for conversion of alcohols into -substituted amides using envirocat EPZG as a catalyst.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 725-38-2
Molecular Formula C14H12FNO
Molecular Weight (g/mol) 229.254
InChI Key XCRBOSJQBAFSMZ-UHFFFAOYSA-N
Synonym benzamide,4-fluoro-n-phenylmethyl, n-benzyl-4-fluoro-benzamide, 4-fluorophenyl-n-benzylcarboxamide
PubChem CID 826977
IUPAC Name N-benzyl-4-fluorobenzamide
SMILES C1=CC=C(C=C1)CNC(=O)C2=CC=C(C=C2)F

Specifications

Quantity 1 g
Formula Weight 229.25
Percent Purity 97%
Chemical Name or Material N-Benzyl-4-fluorobenzamide
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.