Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Benzyl-4-methoxybenzamide, 97%, Thermo Scientific™

Catalog No. AAH5942103
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5942103 1 g
AAH59421MD 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5942103 Supplier Thermo Scientific Chemicals Supplier No. H5942103

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Solubility
Soluble in water.

Notes
Keep container tightly closed. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7465-87-4
Molecular Formula C15H15NO2
Molecular Weight (g/mol) 241.29
MDL Number MFCD00507374
InChI Key GEFZSLMGZREDTO-UHFFFAOYSA-N
Synonym n-benzyl-4-methoxy-benzamide, aronis25526, 4-methoxyphenyl-n-benzylcarboxamide
PubChem CID 346035
IUPAC Name N-benzyl-4-methoxybenzamide
SMILES COC1=CC=C(C=C1)C(=O)NCC2=CC=CC=C2

Specifications

Quantity 1 g
Solubility Information Soluble in water.
Formula Weight 241.29
Percent Purity 97%
Physical Form Solid
Chemical Name or Material N-Benzyl-4-methoxybenzamide
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.