Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Benzylacetamide 98.0+%, TCI America™
SDP

Catalog No. B040425G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B040425G 25 g
B0404500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B040425G Supplier TCI America Supplier No. B040425G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 588-46-5
Molecular Formula C9H11NO
Molecular Weight (g/mol) 149.193
MDL Number MFCD00059204
InChI Key UZJLYRRDVFWSGA-UHFFFAOYSA-N
Synonym N-Acetylbenzylamine
PubChem CID 11500
IUPAC Name N-benzylacetamide
SMILES CC(=O)NCC1=CC=CC=C1

Specifications

Melting Point 61°C
Color White-Yellow
Boiling Point 157°C
Quantity 25 g
Formula Weight 149.19
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material N-Benzylacetamide
EINECSNumber (3)-0373&(9)-0024
RTECSNumber AB4546300
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.