Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Benzyldiethanolamine 98.0+%, TCI America™
SDP

Catalog No. B50811G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B50811G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B50811G Supplier TCI America Supplier No. B50811G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 101-32-6
Molecular Formula C11H17NO2
Molecular Weight (g/mol) 195.262
MDL Number MFCD00020587
InChI Key MIZIOHLLYXVEHJ-UHFFFAOYSA-N
Synonym 2,2′C-(Benzylimino)diethanol, N,N-Bis(2-hydroxyethyl)benzylamine
PubChem CID 7553
IUPAC Name 2-[benzyl(2-hydroxyethyl)amino]ethanol
SMILES C1=CC=C(C=C1)CN(CCO)CCO

Specifications

Boiling Point 226°C
Quantity 1 g
Formula Weight 195.26
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material N-Benzyldiethanolamine
RTECSNumber KJ7538000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.