Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Benzylformamide 98.0+%, TCI America™
SDP

Catalog No. B218425G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B218425G 25 g
B21845G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B218425G Supplier TCI America Supplier No. B218425G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 6343-54-0
Molecular Formula C8H9NO
Molecular Weight (g/mol) 135.166
MDL Number MFCD00003281
InChI Key IIBOGKHTXBPGEI-UHFFFAOYSA-N
Synonym benzyl formamide, n-phenylmethyl formamide, n-benzyl-formamide, unii-l363d92elk, formamide, n-phenylmethyl, formamide,n-phenylmethyl, chembl46293, n-benzyl formamide, n-benzylcarboxamide, n-formylbenzylamine
PubChem CID 80654
ChEBI CHEBI:41117
IUPAC Name N-benzylformamide
SMILES C1=CC=C(C=C1)CNC=O

Specifications

Melting Point 62°C
Color White-Yellow
Quantity 25 g
Formula Weight 135.17
Percent Purity ≥98.0% (GC,N)
Physical Form Crystalline Powder
Chemical Name or Material N-Benzylformamide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.