Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Benzylglycine Hydrochloride 98.0+%, TCI America™
SDP

Catalog No. B307525G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B307525G 25 g
B30755G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B307525G Supplier TCI America Supplier No. B307525G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 7689-50-1
Molecular Formula C9H12ClNO2
Molecular Weight (g/mol) 201.65
MDL Number MFCD00156934
InChI Key BUZJPENZWLUHJD-UHFFFAOYSA-N
Synonym n-benzylglycine hydrochloride, benzylaminoacetic acid hydrochloride, 2-benzylamino acetic acid hydrochloride, bzl-gly-oh.hcl, n-benzylglycine hcl, bz-gly-oh.hcl, n-benzyl glycine hydrochloride, bzl-gly-oh inverted exclamation mark currencyhcl, benzylamino acetic acid hydrochloride, glycine, n-phenylmethyl-, hydrochloride
PubChem CID 2756624
IUPAC Name 2-(benzylamino)acetic acid;hydrochloride
SMILES C1=CC=C(C=C1)CNCC(=O)O.Cl

Specifications

Melting Point 228°C
Color White-Yellow
Quantity 25 g
Formula Weight 201.65
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material N-Benzylglycine Hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.