Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Benzylhydroxylamine Hydrochloride 98.0+%, TCI America™
SDP

Catalog No. B407325G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B407325G 25 g
B40735G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B407325G Supplier TCI America Supplier No. B407325G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 29601-98-7
Molecular Formula C7H10ClNO
Molecular Weight (g/mol) 159.613
MDL Number MFCD00043377
InChI Key YSNXOQGDHGUKCZ-UHFFFAOYSA-N
Synonym n-benzylhydroxylamine hydrochloride, n-benzylhydroxylamine hcl, n-benzyl-n-hydroxylamine hydrochloride, benzenemethanamine, n-hydroxy-, hydrochloride, n-benzylhydroxyamine hydrochloride, bnnhoh*hydrochloride;, acmc-1cr2p, ksc493k8t, benzylhydroxyamine hydrochloride, n-benzylhydroxylamne hydrochlorde
PubChem CID 11332622
IUPAC Name N-benzylhydroxylamine;hydrochloride
SMILES C1=CC=C(C=C1)CNO.Cl

Specifications

Melting Point 106°C
Color White
Quantity 25 g
Formula Weight 159.61
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material N-Benzylhydroxylamine Hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.