Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Benzylidenebenzenesulfonamide 98.0+%, TCI America™
SDP

Catalog No. B220625G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B220625G 25 g
B22065G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B220625G Supplier TCI America Supplier No. B220625G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 13909-34-7
Molecular Formula C13H11NO2S
Molecular Weight (g/mol) 245.30
MDL Number MFCD00011587
InChI Key MPNRJEPBAYEQBY-UHFFFAOYSA-N
PubChem CID 9602006
IUPAC Name N-(phenylmethylidene)benzenesulfonamide
SMILES O=S(=O)(N=CC1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 86°C
Quantity 25 g
Formula Weight 245.30
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material N-Benzylidenebenzenesulfonamide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.