Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Benzylnortropinone 95.0+%, TCI America™
SDP

Catalog No. B40581G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B40581G 1 g
B40585G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B40581G Supplier TCI America Supplier No. B40581G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 28957-72-4
Molecular Formula C14H17NO
Molecular Weight (g/mol) 215.30
MDL Number MFCD00151640
InChI Key RSUHKGOVXMXCND-UHFFFAOYNA-N
Synonym 8-benzyl-8-azabicyclo 3.2.1 octan-3-one, n-benzylnortropinone, n-benzyltropinone, 8-benzyl-8-aza-bicyclo 3.2.1 octan-3-one, 8-phenylmethyl-8-azabicyclo 3.2.1 octan-3-one, 8-azabicyclo 3.2.1 octan-3-one, 8-phenylmethyl, n-benzyl tropinone, 8-benzyl-8-azabicyclo 3.2.1 octan-3-on, n-benzyl-8-azabicyclo 3.2.1 octan-3-one
PubChem CID 119846
IUPAC Name 8-benzyl-8-azabicyclo[3.2.1]octan-3-one
SMILES O=C1CC2CCC(C1)N2CC1=CC=CC=C1

Specifications

Boiling Point 124°C
Quantity 1 g
Formula Weight 215.30
Percent Purity ≥95.0% (GC,T)
Physical Form Liquid
Chemical Name or Material N-Benzylnortropinone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.