Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Benzyloxycarbonyl-2-methylalanine, 98%, Thermo Scientific Chemicals

Catalog No. AAH6311903
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6311903 1 g
AAH6311906 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6311903 Supplier Thermo Scientific Chemicals Supplier No. H6311903
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 15030-72-5
Molecular Formula C12H15NO4
Molecular Weight (g/mol) 237.255
MDL Number MFCD00004190
InChI Key QKVCSJBBYNYZNM-UHFFFAOYSA-N
Synonym z-aib-oh, n-cbz-2-methylalanine, 2-benzyloxy carbonyl amino-2-methylpropanoic acid, n-cbz-2-amino-2-methylpropanoic acid, z-2-aminoisobutyric acid, z-2-methylalanine, n-carbobenzyloxy-2-methylalanine, 2-carbobenzoxyamino isobutyric acid, n-benzyloxy carbonyl-2-methylalanine, alanine, 2-methyl-n-phenylmethoxy carbonyl
PubChem CID 294936
IUPAC Name 2-methyl-2-(phenylmethoxycarbonylamino)propanoic acid
SMILES CC(C)(C(=O)O)NC(=O)OCC1=CC=CC=C1

Specifications

Melting Point 65°C to 69°C (decomposition)
Color White
Beilstein 2218009
Quantity 1 g
Formula Weight 237.25
Percent Purity 98%
Physical Form Powder
Chemical Name or Material N-Benzyloxycarbonyl-2-methylalanine
TSCA No
storageNote1 2° to 8°C
Recommended Storage Keep cold

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.