Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Benzyloxycarbonyl-4-trans-hydroxy-L-proline methyl ester, 98%, Thermo Scientific Chemicals

Catalog No. AAH6399303
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6399303 1 g
AAH6399306 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6399303 Supplier Thermo Scientific Chemicals Supplier No. H6399303

Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please to view product availability in your area.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 64187-48-0
Molecular Formula C14H17NO5
Molecular Weight (g/mol) 279.29
MDL Number MFCD00055851,MFCD09954933,MFCD09954934
InChI Key VVKAGQHUUDRPOI-UHFFFAOYNA-N
Synonym z-hyp-ome, 2s,4r-1-benzyl 2-methyl 4-hydroxypyrrolidine-1,2-dicarboxylate, n-cbz-l-4-hydroxyproline methyl ester, 1-benzyl 2-methyl 2s,4r-4-hydroxypyrrolidine-1,2-dicarboxylate, n-cbz-trans-4-hydroxy-l-proline methyl ester, trans-n-cbz-4-hydroxy-l-proline methyl ester, l-zhprome, z-l-hyp-ome, z-hydroxy-l-proline methyl ester, 1,2-pyrrolidinedicarboxylicacid, 4-hydroxy-, 2-methyl 1-phenylmethyl ester, 2s,4r
PubChem CID 688411
IUPAC Name 1-O-benzyl 2-O-methyl (2S,4R)-4-hydroxypyrrolidine-1,2-dicarboxylate
SMILES COC(=O)C1CC(O)CN1C(=O)OCC1=CC=CC=C1

Specifications

Color Yellow
Quantity 1 g
Formula Weight 279.29
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material N-Benzyloxycarbonyl-4-trans-hydroxy-L-proline methyl ester
TSCA No
storageNote1 2° to 8°C
Recommended Storage Keep cold

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.