Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Benzyloxycarbonyl-D-phenylalaninol, 98%

Catalog No. AAH2781903
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2781903 1 g
AAH2781906 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH2781903 Supplier Thermo Scientific Chemicals Supplier No. H2781903
Add to Cart

CAS: 58917-85-4 | C17H19NO3 | 285.34 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 58917-85-4
Molecular Formula C17H19NO3
Molecular Weight (g/mol) 285.34
MDL Number MFCD00191193
InChI Key WPOFMMJJCPZPAO-MRXNPFEDSA-N
Synonym cbz-d-phenylalaninol, z-d-phenylalaninol, n-carbobenzoxy-d-phenylalaninol, r-+-2-cbz-amino-3-phenyl-1-propanol, r-+-2-benzyloxycarbonylamino-3-phenyl-1-propanol, n-carbobenzyloxy-d-phenylalaninol, r-+-2-carbobenzyloxyamino-3-phenyl-1-propanol, benzyl n-2r-1-hydroxy-3-phenylpropan-2-yl carbamate, r-2-z-amino-3-phenyl-1-propanol, benzyl 2r-1-hydroxy-3-phenylpropan-2-yl carbamate
PubChem CID 736170
IUPAC Name benzyl N-[(2R)-1-hydroxy-3-phenylpropan-2-yl]carbamate
SMILES OC[C@@H](CC1=CC=CC=C1)NC(=O)OCC1=CC=CC=C1

Specifications

Melting Point 92°C to 95°C
Quantity 1 g
Optical Rotation +42° (c=1 in Methanol)
Formula Weight 285.35
Percent Purity 98%
Chemical Name or Material N-Benzyloxycarbonyl-D-phenylalaninol
Hazard Category H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.