Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Benzyloxycarbonyl-D-proline, 98+%

Catalog No. AAB2500514
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2500514 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAB2500514 Supplier Thermo Scientific Chemicals Supplier No. B2500514

missing translation for 'validMainframeMsg'

CAS: 6404-31-5 | C13H15NO4 | 249.266 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6404-31-5
Molecular Formula C13H15NO4
Molecular Weight (g/mol) 249.266
MDL Number MFCD00063228
InChI Key JXGVXCZADZNAMJ-LLVKDONJSA-N
Synonym n-benzyloxycarbonyl-d-proline, z-d-pro-oh, +-z-d-proline, z-d-proline, r-n-benzyloxycarbonylproline, n-cbz-d-proline, n-carbobenzoxy-d-proline, 1-benzyloxy carbonyl-d-proline, r-1-benzyloxy carbonyl pyrrolidine-2-carboxylic acid, 2r-1-benzyloxy carbonyl pyrrolidine-2-carboxylic acid
PubChem CID 111208
IUPAC Name (2R)-1-phenylmethoxycarbonylpyrrolidine-2-carboxylic acid
SMILES C1CC(N(C1)C(=O)OCC2=CC=CC=C2)C(=O)O

Specifications

Melting Point 76°C to 78°C
Quantity 25 g
Beilstein 485188
Optical Rotation +40° (c=2 in Ethanol)
Formula Weight 249.27
Percent Purity ≥98%
Chemical Name or Material N-Benzyloxycarbonyl-D-proline
EINECSNumber 229-021-5
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.