Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Benzyloxycarbonyl-O-benzyl-L-tyrosine, 95%, Thermo Scientific™

Catalog No. AAH6348703
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6348703 1 g
AAH6348706 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6348703 Supplier Thermo Scientific Chemicals Supplier No. H6348703

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 16677-29-5
Molecular Formula C24H23NO5
Molecular Weight (g/mol) 405.45
MDL Number MFCD00037819
InChI Key IPAODWFPTVIUSZ-ANBDAQEENA-N
Synonym z-tyr bzl-oh, z-o-benzyl-l-tyrosine, cbz-tyr bzl-oh, cbz-o-benzyl-l-tyrosine, s-2-benzyloxy carbonyl amino-3-4-benzyloxy phenyl propanoic acid, l-tyrosine, n-phenylmethoxy carbonyl-o-phenylmethyl, 2s-2-benzyloxy carbonyl amino-3-4-benzyloxy phenyl propanoic acid, z-tyr bzl, cbz-o-benzyl-l-tyr, pubchem19061
PubChem CID 47600
IUPAC Name (2S)-2-(phenylmethoxycarbonylamino)-3-(4-phenylmethoxyphenyl)propanoic acid
SMILES OC(=O)[C@H](CC1=CC=C(OCC2=CC=CC=C2)C=C1)NC(=O)OCC1=CC=CC=C1

Specifications

Beilstein 3178475
Quantity 1 g
Formula Weight 405.44
Percent Purity 95%
Physical Form Solid
Chemical Name or Material N-Benzyloxycarbonyl-O-benzyl-L-tyrosine
TSCA No
storageNote1 2° to 8°C
Recommended Storage Keep cold
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.