Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Benzyloxycarbonyl-O-tert-butyl-L-tyrosine, 98%

Catalog No. AAH6676214
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6676214 25 g
AAH6676206 5 g
AAH6676222 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH6676214 Supplier Thermo Scientific Chemicals Supplier No. H6676214
Add to Cart

CAS: 5545-54-0 | C21H25NO5 | 371.43 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5545-54-0
Molecular Formula C21H25NO5
Molecular Weight (g/mol) 371.43
MDL Number MFCD00065701
InChI Key YKVBQSGNGCKQSV-UHFFFAOYNA-N
Synonym z-tyr tbu-oh, s-2-benzyloxy carbonyl amino-3-4-tert-butoxy phenyl propanoic acid, n-cbz-o-tert-butyl-l-tyrosine, l-tyrosine, o-1,1-dimethylethyl-n-phenylmethoxy carbonyl, 2s-2-benzyloxy carbonyl amino-3-4-tert-butoxy phenyl propanoic acid, z-tyr but-oh, z-l-tyr tbu-oh, z-tyr t-bu-oh, n-benzyloxycarbonyl-o-t-butyl-l-tyrosine, o-tert-butyl-n-benzyloxycarbonyl tyrosine
PubChem CID 978520
IUPAC Name 2-{[(benzyloxy)carbonyl]amino}-3-[4-(tert-butoxy)phenyl]propanoic acid
SMILES CC(C)(C)OC1=CC=C(CC(NC(=O)OCC2=CC=CC=C2)C(O)=O)C=C1

Specifications

Color White
Beilstein 2781067
Quantity 25 g
Formula Weight 371.43
Percent Purity 98%
Physical Form Powder
Chemical Name or Material N-Benzyloxycarbonyl-O-tert-butyl-L-tyrosine
TSCA No
storageNote1 2° to 8°C
Recommended Storage Keep cold

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.