Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Boc-2-trifluoromethyl-D-phenylalanine, 95%, Thermo Scientific™

Catalog No. AAH6234806
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6234806 5 g
AAH62348MD 250 mg
AAH6234803 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH6234806 Supplier Thermo Scientific Chemicals Supplier No. H6234806

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 346694-78-8
Molecular Formula C15H18F3NO4
Molecular Weight (g/mol) 333.31
MDL Number MFCD01862945,MFCD01862946
InChI Key XKMOOODKNPYTEE-UHFFFAOYNA-N
Synonym boc-d-2-trifluoromethylphenylalanine, n-boc-2-trifluoromethyl-d-phenylalanine, boc-2-trifluoromethyl-d-phenylalanine, r-2-tert-butoxycarbonyl amino-3-2-trifluoromethyl phenyl propanoic acid, boc-d-2-trifluoromethylphe, boc-d-phe 2-cf3-oh, 2r-2-tert-butoxycarbonyl amino-3-2-trifluoromethyl phenyl propanoic acid, boc-d-2-trifluoromethyl-phe-oh
PubChem CID 7010037
SMILES CC(C)(C)OC(=O)NC(CC1=CC=CC=C1C(F)(F)F)C(O)=O

Specifications

IUPAC Name 2-{[(tert-butoxy)carbonyl]amino}-3-[2-(trifluoromethyl)phenyl]propanoic acid
Quantity 5 g
Formula Weight 333.3
Percent Purity 95%
Physical Form Solid
Chemical Name or Material N-Boc-2-trifluoromethyl-D-phenylalanine
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.