Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Boc-3-(2-furyl)-L-alanine dicyclohexylammonium salt, 98%, Thermo Scientific™

Catalog No. AAH6225803
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6225803 1 g
AAH62258MD 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6225803 Supplier Thermo Scientific Chemicals Supplier No. H6225803

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 331730-08-6
Molecular Formula C24H40N2O5
Molecular Weight (g/mol) 436.593
MDL Number MFCD01860894
InChI Key QYEOQPFAYJEDFE-FVGYRXGTSA-N
Synonym dicyclohexylamine s-2-tert-butoxycarbonyl amino-3-furan-2-yl propanoate, boc-l-2-furylalanine-dcha, 2s-2-tert-butoxycarbonyl amino-3-furan-2-yl propanoic acid; dicha, dicycloammonium fur-2-yl-l-alanine, n-boc protected, boc-l-2-furylalanine.dcha, boc-beta-2-furyl-ala-ohcha, boc-l-2-furylalanine dicyclohexylamine salt, boc-3-2-furyl-l-alanine dicyclohexylamine, boc-3-2-furyl-l-alaninedicyclohexylaminesalt, boc-beta-2-furyl-ala-oh dicyclohexylammonium salt
PubChem CID 16211161
IUPAC Name N-cyclohexylcyclohexanamine;(2S)-3-(furan-2-yl)-2-[(2-methylpropan-2-yl)oxycarbonylamino]propanoic acid
SMILES CC(C)(C)OC(=O)NC(CC1=CC=CO1)C(=O)O.C1CCC(CC1)NC2CCCCC2

Specifications

Color White
Quantity 1 g
Formula Weight 436.58
Percent Purity 98%
Physical Form Powder
Chemical Name or Material N-Boc-3-(2-furyl)-L-alanine dicyclohexylammonium salt
TSCA No
storageNote1 2° to 8°C
Recommended Storage Keep cold
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.