Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Boc-3-(4-biphenylyl)-D-alanine, 95%, Thermo Scientific Chemicals

Catalog No. AAH63494MD
Change view
Click to view available options
Quantity:
250 mg
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH63494MD 250 mg
AAH6349406 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH63494MD Supplier Thermo Scientific Chemicals Supplier No. H63494MD

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

View Alternate Product

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 128779-47-5
Molecular Formula C20H23NO4
Molecular Weight (g/mol) 341.41
MDL Number MFCD00191185
InChI Key NBVVKAUSAGHTSU-UHFFFAOYNA-N
Synonym boc-d-4,4'-biphenylalanine, boc-d-4,4-biphenylalanine, boc-3-4-biphenylyl-d-alanine, r-3-1,1'-biphenyl-4-yl-2-tert-butoxycarbonyl amino propanoic acid, boc-d-bip-oh, boc-d-4-phenylphenylalanine, boc-4-biphenyl-d-ala, boc-4-phenyl-d-phe-oh, boc-d-bph-oh, boc-d-bip 4,4'-oh
PubChem CID 7019133
SMILES CC(C)(C)OC(=O)NC(CC1=CC=C(C=C1)C1=CC=CC=C1)C(O)=O

Specifications

IUPAC Name 3-{[1,1'-biphenyl]-4-yl}-2-{[(tert-butoxy)carbonyl]amino}propanoic acid
Quantity 250 mg
Formula Weight 341.41
Percent Purity 95%
Physical Form Solid
Chemical Name or Material N-Boc-3-(4-biphenylyl)-D-alanine
TSCA No
storageNote1 -30° to -10°C
Recommended Storage Store at -20°C

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.