Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Boc-3-(4-pyridyl)-D-alanine, 95%

Catalog No. AAH62215MD
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH62215MD 250 mg
AAH6221503 1 g
AAH6221506 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH62215MD Supplier Thermo Scientific Chemicals Supplier No. H62215MD

CAS: 37535-58-3 | C13H18N2O4 | 266.297 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 37535-58-3
Molecular Formula C13H18N2O4
Molecular Weight (g/mol) 266.297
MDL Number MFCD00672529
InChI Key FNYWDMKESUACOU-SNVBAGLBSA-N
Synonym boc-d-4-pyridylalanine, boc-d-4-pal-oh, r-2-tert-butoxycarbonyl amino-3-pyridin-4-yl propanoic acid, boc-3-4-pyridyl-d-alanine, boc-d-4-pyridylala, boc-4'-pyridyl-d-ala, boc-3-4-pyridyl-d-ala-oh, n-tert-butoxycarbonyl-4-pyridyl-d-alanine, boc-d-3-4-pyridyl-alanine, r-2-tert-butoxycarbonylamino-3-pyridin-4-yl-propionic acid
PubChem CID 2734503
IUPAC Name (2R)-2-[(2-methylpropan-2-yl)oxycarbonylamino]-3-pyridin-4-ylpropanoic acid
SMILES CC(C)(C)OC(=O)NC(CC1=CC=NC=C1)C(=O)O

Specifications

Melting Point 223°C to 229°C
Color White
Beilstein 4195478
Quantity 250 mg
Formula Weight 266.29
Sensitivity Air and moisture sensitive
Percent Purity 95%
Physical Form Powder
Chemical Name or Material N-Boc-3-(4-pyridyl)-D-alanine
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
storageNote1 2° to 8°C
Recommended Storage Keep cold; Store under Argon

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.