Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-BOC-4-methylene-L-proline, 97%

Catalog No. AC449260010
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC449260010 1 g
AC449260050 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC449260010 Supplier Thermo Scientific Chemicals Supplier No. 449260010
Add to Cart

CAS: 84348-38-9 | C11H17NO4 | 227.26 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 84348-38-9
Molecular Formula C11H17NO4
Molecular Weight (g/mol) 227.26
MDL Number MFCD01861787
InChI Key ULLGRIBXGPATMA-UHFFFAOYNA-N
Synonym n-boc-4-methylene-l-proline, s-1-tert-butoxycarbonyl-4-methylenepyrrolidine-2-carboxylic acid, 1-boc-s-4-methylenepyrrolidine-2-carboxylic acid, s-4-methylene-pyrrolidine-1,2-dicarboxylic acid 1-tert-butyl ester, 2s-1-tert-butoxycarbonyl-4-methylidenepyrrolidine-2-carboxylic acid, 1,2-pyrrolidinedicarboxylicacid, 4-methylene-, 1-1,1-dimethylethyl ester, 2s, n-boc-4-methylene-l-prolne, n-t-boc-4-methylene-l-proline, 1-tert-butoxycarbonyl-4-methyleneproline
PubChem CID 21727417
IUPAC Name (2S)-4-methylidene-1-[(2-methylpropan-2-yl)oxycarbonyl]pyrrolidine-2-carboxylic acid
SMILES CC(C)(C)OC(=O)N1CC(=C)CC1C(O)=O

Specifications

Melting Point 110.0°C to 115.0°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass Bottle
Quantity 1 g
Solubility Information Solubility in water: insoluble.
Formula Weight 227.26
Percent Purity 97%
Chemical Name or Material N-BOC-4-methylene-L-proline
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.